Learn More
Medchemexpress LLC HY-112504 5mg Medchemexpress, INCB054329 CAS:1628607-64-6 Purity:>98%
SDP
Catalog No. 501969099
Encompass_Preferred
Medchemexpress, HY-112504 5mg INCB054329 CAS:1628607-64-6 INCB054329 is a potent BET inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9099 Supplier Medchemexpress LLC Supplier No. HY1125045MG
Medchemexpress, HY-112504 5mg INCB054329 CAS:1628607-64-6 INCB054329 is a potent BET inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.