Learn More
Selleck Chemical LLC INCB059872 dihydrochloride
Supplier: Selleck Chemical LLC E1899100MG
INCB059872 dihydrochloride is a potent selective and irreversible inhibitor of Lysine-Specific Demethylase 1(LSD1) It exhibits potential antineoplastic activity and can be used in acute myeloid leukemia (AML) research