Learn More
Medchemexpress LLC INCB059872 dihydrochloride | 3062683-32-0 | 97.9% | C23H34N2O3 • 2HCl | 50 MG
SDP
Supplier:  Medchemexpress LLC HY141677A50MG
Encompass_Preferred
INCB059872 dihydrochloride is a potent, selective, and irreversible inhibitor of Lysine-Specific Demethylase 1 (LSD1). It exhibits potential antineoplastic activity and can be used in acute myeloid leukemia (AML) research. It has been shown to decrease proliferation in THP-1 acute monocytic and MV4-11 acute myeloid leukemia cells, and to increase the levels of CD11B in THP-1 cells when used at a concentration of 100 nM.
  • Potent, selective, and irreversible LSD1 inhibitor
  • Potential antineoplastic activity
  • Useful in acute myeloid leukemia (AML) research
  • Decreases proliferation in acute monocytic and myeloid leukemia cells
  • Increases levels of CD11B in THP-1 cells
  • Cited in Nature Medicine
Catalog No. 50-003-53409