Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC INCB059872 dihydrochloride | 3062683-32-0 | 99.2% | 459.45 g·mol⁻¹ | C23H36Cl2N2O3 | 5 MG
SDP

Catalog No. 5000221350
Encompass_Preferred

This compound is the dihydrochloride salt of a potent, orally active, selective and irreversible lysine-specific demethylase 1 (LSD1/KDM1A) inhibitor intended for in vitro and preclinical cancer research, including studies of myeloid leukemia.

  • Potent, selective LSD1 (KDM1A) inhibitory activity suitable for epigenetic research.
  • High purity: 99.18% by supplier analysis.
  • White to light yellow solid appearance for reliable handling.
  • Molecular formula C23H36Cl2N2O3 and molecular weight 459.45 g·mol⁻¹.
  • Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Suitable for in vitro assays and preclinical studies related to leukemia and other cancers.
  • Supplied in small research quantities for laboratory use.

Catalog No. 50-002-21350 Supplier Medchemexpress LLC Supplier No. HY141677A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This compound is the dihydrochloride salt of a potent, orally active, selective and irreversible lysine-specific demethylase 1 (LSD1/KDM1A) inhibitor intended for in vitro and preclinical cancer research, including studies of myeloid leukemia.

  • Potent, selective LSD1 (KDM1A) inhibitory activity suitable for epigenetic research.
  • High purity: 99.18% by supplier analysis.
  • White to light yellow solid appearance for reliable handling.
  • Molecular formula C23H36Cl2N2O3 and molecular weight 459.45 g·mol⁻¹.
  • Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Suitable for in vitro assays and preclinical studies related to leukemia and other cancers.
  • Supplied in small research quantities for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.