Learn More
Medchemexpress LLC Incb059872 dihydrochloride | 3062683-32-0 | 99.2% | C23H36Cl2N2O3 | 10MG
SDP
Catalog No. 5000260759
Encompass_Preferred
INCB059872 dihydrochloride is the dihydrochloride salt of a potent, orally active, selective, and irreversible inhibitor of lysine-specific demethylase 1 (LSD1). Supplied for laboratory research use only, the compound is provided at high purity and has been used in preclinical studies of myeloid leukemia and epigenetic regulation.
  • Potent, selective LSD1 inhibition.
  • Orally active in preclinical models.
  • High chemical purity (99.18%).
  • Dihydrochloride salt form for improved stability.
  • Intended for laboratory research use only.
Catalog No. 50-002-60759 Supplier Medchemexpress LLC Supplier No. HY141677A10MG
INCB059872 dihydrochloride is the dihydrochloride salt of a potent, orally active, selective, and irreversible inhibitor of lysine-specific demethylase 1 (LSD1). Supplied for laboratory research use only, the compound is provided at high purity and has been used in preclinical studies of myeloid leukemia and epigenetic regulation.
  • Potent, selective LSD1 inhibition.
  • Orally active in preclinical models.
  • High chemical purity (99.18%).
  • Dihydrochloride salt form for improved stability.
  • Intended for laboratory research use only.