Learn More
Selleck Chemical LLC INCB059872 dihydrochloride
SDP
Catalog No. 503134778
Encompass_Preferred
INCB059872 dihydrochloride is a potent selective and irreversible inhibitor of Lysine-Specific Demethylase 1(LSD1) It exhibits potential antineoplastic activity and can be used in acute myeloid leukemia (AML) research
Catalog No. 50-313-4778 Supplier Selleck Chemical LLC Supplier No. E18995MG
Add to Cart
Add to Cart
INCB059872 dihydrochloride is a potent selective and irreversible inhibitor of Lysine-Specific Demethylase 1(LSD1) It exhibits potential antineoplastic activity and can be used in acute myeloid leukemia (AML) research