Learn More
Apexbio Technology LLC INCB28060, 50mg. Cas: 1029712-80-8 MFCD: MFCD18633285. C-Met inhibitor,ATP-competitive and novel
SDP
Supplier:  Apexbio Technology LLC A844850
Encompass_Preferred
INCB28060, also known as Capmatinib and INC280, is a novel and ATP-competitive inhibitor of c-Met with IC50 of 0.13 nM.c-Met, also named as hepatocyte growth factor receptor (HGFR) is a receptor tyrosine kinase that is essential for embryonic development and would healing. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5145