Learn More
Medchemexpress LLC HY-50674 100mg , INCB3344 CAS:1262238-11-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY50674100MG
Encompass_Preferred
HY-50674 100mg INCB3344 CAS: 1262238-11-8 INCB3344 is a potent, selective and orally bioavailable CCR2 antagonist with IC50 values of 5.1 nM (hCCR2) and 9.5 nM (mCCR2) in binding antagonism and 3.8 nM (hCCR2) and 7.8 nM (mCCR2) in antagonism of chemotaxis activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1075