Learn More
Medchemexpress LLC INCB3344 R-isomer | 1262238-11-8 | 98.0% | C29H34F3N3O6 | 1 ML
SDP
Catalog No. 5000359286
Encompass_Preferred
INCB3344 R-isomer is the R-isomer of INCB3344, a potent, selective, and orally bioavailable CCR2 antagonist. It demonstrates high binding antagonism and chemotaxis activity against human and murine CCR2. This compound is for research use only and exhibits high selectivity for hCCR2 over a broad panel of other targets.
  • Potent and selective CCR2 antagonist
  • Orally bioavailable
  • Active against human and murine CCR2
  • High selectivity for hCCR2 over many other ion channels, transporters, and GPCRs
  • Prevents vascular CCR2 expression changes in specific models
Catalog No. 50-003-59286 Supplier Medchemexpress LLC Supplier No. HY50674A10MM/1ML
INCB3344 R-isomer is the R-isomer of INCB3344, a potent, selective, and orally bioavailable CCR2 antagonist. It demonstrates high binding antagonism and chemotaxis activity against human and murine CCR2. This compound is for research use only and exhibits high selectivity for hCCR2 over a broad panel of other targets.
  • Potent and selective CCR2 antagonist
  • Orally bioavailable
  • Active against human and murine CCR2
  • High selectivity for hCCR2 over many other ion channels, transporters, and GPCRs
  • Prevents vascular CCR2 expression changes in specific models