Learn More
Eurofins Discoverx InCELL Detection Kit, 2,000 (100 plates), 40,000 dp (384-well). (96-0079L)
Catalog No. 502004543
Encompass
The InCELL Detection Kit is a detection kit intended for use with InCELL Hunter or Pulse cell lines, allowing measurement of target-engagement based changes in protein stability. This kit contains a lysis buffer and exogenous EA as well as EFC substrate to allow for detection of ProLabel-tagged protein levels in the assayed cells. This kit contains all components required to assay 100 plates (96- or 384-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-96-0079l.
Catalog No. 50-200-4543 Supplier Eurofins Discoverx Supplier No. 960079L
Add to Cart
Add to Cart
Request a Quote
The InCELL Detection Kit is a detection kit intended for use with InCELL Hunter or Pulse cell lines, allowing measurement of target-engagement based changes in protein stability. This kit contains a lysis buffer and exogenous EA as well as EFC substrate to allow for detection of ProLabel-tagged protein levels in the assayed cells. This kit contains all components required to assay 100 plates (96- or 384-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-96-0079l.