Learn More
Eurofins Discoverx InCELL Detection Kit, 216 (16 plates), 1,600/6,400 dp (96-well/384-well). (96-0079)
Supplier:  Eurofins Discoverx 960079
Encompass
The InCELL Detection Kit is a detection kit intended for use with InCELL Hunter or Pulse cell lines, allowing measurement of target-engagement based changes in protein stability. This kit contains a lysis buffer and exogenous EA as well as EFC substrate to allow for detection of ProLabel-tagged protein levels in the assayed cells. This kit contains all components required to assay 16 plates (96- or 384-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-96-0079.
Catalog No. 50-200-4540