Learn More
AdipoGen Incensol (1 mg)
SDP
Supplier:  AdipoGen AGCN20482M001
Encompass_Preferred
Chemical. CAS: 22419-74-5. Formula: C20H34O2. MW: 306.5. Isolated from Boswellia papyrifera. Component of the frankincense essential oil. Activator of transient receptor potential vanilloid 3 (TRPV3), a heat-sensitive ion channel involved in skin heat sensitivity, thermoregulation and neurological activities. More powerful TRPV3 agonist than incensol acetate (Prod. No. AG-CN2-0484). Potent inhibitor of STAT3. Showed inhibitory activity on IL-6-induced STAT3 activation. Shown to have COX-1 and COX-2 inhibitory activity and cytotoxic activity against selected cancer cell lines.
Catalog No. 50-205-3665