Learn More
Medchemexpress LLC Incensole | 22419-74-5 | 5 MG
SDP
Catalog No. 5000418765
Encompass_Preferred
Incensole is a 14-membered diterpenoid isolated from frankincense essential oils and resins. It exhibits anti-inflammatory and anti-depression activities by activating ion channels in the brain to alleviate anxiety or depression.
  • Activator of TRPV3 channels
  • Demonstrated anti-inflammatory activity
  • Exhibits anti-depression activity
  • Activates ion channels in the brain
  • Isolated from frankincense
Catalog No. 50-004-18765 Supplier Medchemexpress LLC Supplier No. HYN40975MG
Incensole is a 14-membered diterpenoid isolated from frankincense essential oils and resins. It exhibits anti-inflammatory and anti-depression activities by activating ion channels in the brain to alleviate anxiety or depression.
  • Activator of TRPV3 channels
  • Demonstrated anti-inflammatory activity
  • Exhibits anti-depression activity
  • Activates ion channels in the brain
  • Isolated from frankincense