Learn More
Medchemexpress LLC HY-N4098 5mg Medchemexpress, Incensole Acetate CAS:34701-53-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN40985MG
Encompass_Preferred
Medchemexpress, HY-N4098 5mg Incensole Acetate CAS:34701-53-6 Incensole acetate is a main constituent of Boswellia carterii resin, has neuroprotective effects against neuronal damage in traumatic and ischemic head injury. Incensole acetate reduces Aβ25–35-triggered apoptosis in hOBNSCs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2923