Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Inclined Plane with Force Mechanism
GREENER_CHOICESCI_ED

Catalog No. S139428 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139428 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139428 Supplier Supertek™ Supplier No. PH10580
Add to Cart
Add to Cart

This Supertek™ Inclined Plane with Force Mechanism apparatus is designed to perform experiments on force and acceleration.

  • This 3.94 ft. aluminum track has a spring based force mechanism mounted permanently at one end.
  • The spring mechanism produces force using a multi position trigger.
  • Determine acceleration speed and distance attained by applying a variable force.
  • The track is mounted on three plastic feet.
  • The apparatus comes with five steel balls.
  • Operating instructions included.

Specifications

Grade 6+
Format Kit
Languages English
Subject Physical Science - Force and Mass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.