Learn More
Medchemexpress LLC Inclisiran sodium (sense strand, no GalNAc) | 00-00-0 | 98.1% | C529H707F12N176O316P43S6 | 10MG
SDP
Catalog No. 5000261342
Encompass_Preferred
SS(no Galnac)-Inclisiran sodium is a single-strand variant of inclisiran supplied as the sodium salt for research use. Inclisiran is a small interfering RNA (siRNA) that reduces PCSK9 expression via RNA interference; the no-GalNAc single-strand form enables studies of strand-specific activity and delivery. The product is provided at high purity and in multiple pack sizes to support in vitro and preclinical workflows.
  • single-strand siRNA without GalNAc for strand-specific studies.
  • siRNA mechanism targeting PCSK9 to reduce transcription.
  • sodium salt form for consistent handling and solubility.
  • high purity suitable for analytical and biological assays.
  • multiple pack sizes to support small- and large-scale experiments.
  • intended for research use only, not for therapeutic use.
  • compatible with in vitro and preclinical lipid and cardiovascular research.
Catalog No. 50-002-61342 Supplier Medchemexpress LLC Supplier No. HY14322010MG
SS(no Galnac)-Inclisiran sodium is a single-strand variant of inclisiran supplied as the sodium salt for research use. Inclisiran is a small interfering RNA (siRNA) that reduces PCSK9 expression via RNA interference; the no-GalNAc single-strand form enables studies of strand-specific activity and delivery. The product is provided at high purity and in multiple pack sizes to support in vitro and preclinical workflows.
  • single-strand siRNA without GalNAc for strand-specific studies.
  • siRNA mechanism targeting PCSK9 to reduce transcription.
  • sodium salt form for consistent handling and solubility.
  • high purity suitable for analytical and biological assays.
  • multiple pack sizes to support small- and large-scale experiments.
  • intended for research use only, not for therapeutic use.
  • compatible with in vitro and preclinical lipid and cardiovascular research.