Learn More
Sigma Aldrich Inconel™ 625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5
Catalog No. 503091902 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
5 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-309-1902 1 Ea.
50-309-1898 5 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-309-1902 Supplier Sigma Aldrich Supplier No. GF357123231EA
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Inconel™ 625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5
Specifications
Quantity 1 Ea.
Synonym NI047905