Learn More
Sigma Aldrich Inconel™ alloy 600 - heat resisting alloy, Ni72Cr16Fe8
Catalog No. 503095600 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
10 Ea.
5 Ea.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-309-5600 5 Ea.
50-309-4355 1 Ea.
50-309-5508 10 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-309-5600 Supplier Sigma Aldrich Supplier No. GF455396135EA
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Inconel™ alloy 600 - heat resisting alloy, Ni72Cr16Fe8
Specifications
Quantity 5 Ea.
Synonym NI027920