Learn More
Anton Paar Increase DLS throughput with the Litesizer Cuvette Sampler, automating up to 80 cuvettes. Kalliope software handles setup and scheduling, freeing users from repetitive tasks. Supports workflows for particle size, zeta potential, concentration, and molecular mas
Catalog No. 5000580525
Encompass_Preferred
Measure liquid and dry samplesMeasure up to 60 samples per runAdd and prioritize samples even during an active runEnsure safety with cobot functionality
Catalog No. 50-005-80525 Supplier Anton Paar Supplier No. 391285
Please contact your sales representative for a quote or further ordering instructions.
Measure liquid and dry samplesMeasure up to 60 samples per runAdd and prioritize samples even during an active runEnsure safety with cobot functionality