Learn More
Perkin Elmer US LLC Spectrum Adulterant Screen
Catalog No. 502016624
Encompass_Preferred
Increasing demand and high-market value of ingredients and food has led to numerous adulteration and fraudulent schemes which undermine the quality attributes and safety of food. Adulterant Screen offers simple method development with a powerful algorithm to ensure detection, identification, and estimation of concentration of materials which have been fraudulently or accidentally introduced into a product. Adulterant Screen is the ideal solution for laboratories needing to test products to verify that they are the stated product, and that they are free from contamination or adulteration. There is no need for extensive software training or methods development. It is particularly useful for, but not limited to, the screening of: Olive oil Cocoa powder Spices Honey Maple syrup Coffee Liquid and powdered milk The Spectrum 10 software is used to collect IR or NIR spectra representing good samples for a product. This consists of multiple spectra representing many sources of variation of th
Catalog No. 50-201-6624 Supplier Perkin Elmer US LLC Supplier No. L1101026
Increasing demand and high-market value of ingredients and food has led to numerous adulteration and fraudulent schemes which undermine the quality attributes and safety of food. Adulterant Screen offers simple method development with a powerful algorithm to ensure detection, identification, and estimation of concentration of materials which have been fraudulently or accidentally introduced into a product. Adulterant Screen is the ideal solution for laboratories needing to test products to verify that they are the stated product, and that they are free from contamination or adulteration. There is no need for extensive software training or methods development. It is particularly useful for, but not limited to, the screening of: Olive oil Cocoa powder Spices Honey Maple syrup Coffee Liquid and powdered milk The Spectrum 10 software is used to collect IR or NIR spectra representing good samples for a product. This consists of multiple spectra representing many sources of variation of th