Learn More
Chemglass Life Sciences Incu-Mixer, 4 Plate, 120V
SDP
Catalog No. 501311293
Encompass
Incu-Mixer, 4 Plate, 120V. The Incu-Mixer MP series is designed to provide an optimal method for vortexing & incubating microplates. Simultaneous heating from both the bottom surface & the lid provide temperature uniformity throughout the sample. The heated lid minimizes evaporation & prevents condensation.2mm orbit ensures thorough mixing in microplates without splashing. Speed, time & temperature are all displayed in real time on the LCD screen while an internal validation system constantly monitors all parameters for accuracy.Designed for application flexibility, the Benchmark Incu-Mixer series is capable of accepting all standard plates up to 40mm high. Independent time, temperature & speed controls allow the mixer to function as three machines in one; a multi-plate incubator, vortexer, or a combination of the two. The Incu-Mixer MP series is ideal for a variety of molecular biology applications including immunochemical reactions, enzyme & protein analysis, & microarray analysis.
Catalog No. 50-131-1293 Supplier Chemglass Life Sciences Supplier No. CLS201804
Incu-Mixer, 4 Plate, 120V. The Incu-Mixer MP series is designed to provide an optimal method for vortexing & incubating microplates. Simultaneous heating from both the bottom surface & the lid provide temperature uniformity throughout the sample. The heated lid minimizes evaporation & prevents condensation.2mm orbit ensures thorough mixing in microplates without splashing. Speed, time & temperature are all displayed in real time on the LCD screen while an internal validation system constantly monitors all parameters for accuracy.Designed for application flexibility, the Benchmark Incu-Mixer series is capable of accepting all standard plates up to 40mm high. Independent time, temperature & speed controls allow the mixer to function as three machines in one; a multi-plate incubator, vortexer, or a combination of the two. The Incu-Mixer MP series is ideal for a variety of molecular biology applications including immunochemical reactions, enzyme & protein analysis, & microarray analysis.