Learn More
Thomas Scientific Incu-Mixer MP2T Heated Plate Vortexer 115-240V (US Plug)
SDP
Catalog No. 501460071
Encompass_Preferred
Optimized for incubated mixing in microplates Heated lid prevents condensation Accepts plates up to 40mm high Independently control time, temp. and speed Available in two-plate and four-plate format The Incu-Mixer MP series by Benchmark Scientific is designed to provide an optimal method for vortexing and incubating microplates. Simultaneous heating from both the bottom surface and the lid provide temperature uniformity throughout the sample. Additionally, the heated lid minimizes evaporation and prevents condensation. A compact, 2mm orbit ensures thorough mixing in microplates without splashing. Speed, time and temperature are all displayed in real time on the large LCD screen while an internal validation system constantly monitors all parameters for accuracy. Designed for application flexibility, the Incu-Mixer series is available in two, and four-plate capacities, both capable of accepting all standard plates up to 40mm high. Independent time, temperature and speed controls allow th
Catalog No. 50-146-0071 Supplier Thomas Scientific Supplier No. 1200F91
Optimized for incubated mixing in microplates Heated lid prevents condensation Accepts plates up to 40mm high Independently control time, temp. and speed Available in two-plate and four-plate format The Incu-Mixer MP series by Benchmark Scientific is designed to provide an optimal method for vortexing and incubating microplates. Simultaneous heating from both the bottom surface and the lid provide temperature uniformity throughout the sample. Additionally, the heated lid minimizes evaporation and prevents condensation. A compact, 2mm orbit ensures thorough mixing in microplates without splashing. Speed, time and temperature are all displayed in real time on the large LCD screen while an internal validation system constantly monitors all parameters for accuracy. Designed for application flexibility, the Incu-Mixer series is available in two, and four-plate capacities, both capable of accepting all standard plates up to 40mm high. Independent time, temperature and speed controls allow th