Learn More
Sigma Aldrich Fine Chemicals Biosciences Incu Mixer MP4 Heated 4 Pl1EA
Catalog No. 502908968
Encompass
The new Incu-Mixer MP series is designed to provide an optimal method for vortexing and incubating microplates. Simultaneous heating from both the bottom surface and the lid provide temperature uniformity throughout the sample. Additionally the heated lid minimizes evaporation and prevents condensation.A compact 2mm orbit ensures thorough mixing in microplates without splashing. Speed time and temperature are all displayed in real time on the large LCD screen while an internal validation system constantly monitors all parameters for accuracy.Designed for application flexibility the Incu-Mixer series is available in two and four-plate capacities both capable of accepting all standard plates up to 40mm high. Independent time temperature and speed controls allow the mixer to function as 3 machines in one a multi-plate incubator vortexer or a combination of the two. The Incu-Mixer MP series is ideal for a wide variety of molecular biology applications including immunochemical reac
Catalog No. 50-290-8968 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BMSH60041EA
The new Incu-Mixer MP series is designed to provide an optimal method for vortexing and incubating microplates. Simultaneous heating from both the bottom surface and the lid provide temperature uniformity throughout the sample. Additionally the heated lid minimizes evaporation and prevents condensation.A compact 2mm orbit ensures thorough mixing in microplates without splashing. Speed time and temperature are all displayed in real time on the large LCD screen while an internal validation system constantly monitors all parameters for accuracy.Designed for application flexibility the Incu-Mixer series is available in two and four-plate capacities both capable of accepting all standard plates up to 40mm high. Independent time temperature and speed controls allow the mixer to function as 3 machines in one a multi-plate incubator vortexer or a combination of the two. The Incu-Mixer MP series is ideal for a wide variety of molecular biology applications including immunochemical reac