Learn More
Chemglass Life Sciences Incu-Shaker CO2 Mini, Shaking, Heating, 120V, with Non-Slip Rubber Mat Platform
SDP
Catalog No. 5000083272
Encompass
Advanced Monitoring Controls Dual Beam IR Sensor Integral Orbital Shaker Stackable Design Stainless Steel Chamber 6-Sided Direct heating. Ideal for the culturing of non-adherent cells, the Incu-Shaker CO2 Mini combines a precision CO2 incubation chamber with an integral orbital shaker, providing the optimum conditions for suspension cell culture. With our unique and proprietary heat distribution technology, the incubator provides the highest levels of temperature accuracy with minimal fluctuation (within 0.1 ° C). The six-side heating system in combination with a ultra-low speed internal fan uniformly spread air throughout the stainless steel chamber without air exchange from the external environment. This minimal air flow design, significantly reduces the concerns of sample to sample contamination and allows for disabling of the fan for applications requiring solely mechanical convection. A powerful, beltless motor drive provides a horizontally circular shaking motion at a consistent
Catalog No. 50-000-83272 Supplier Chemglass Life Sciences Supplier No. CLS254801
Add to Cart
Add to Cart
Request a Quote
Advanced Monitoring Controls Dual Beam IR Sensor Integral Orbital Shaker Stackable Design Stainless Steel Chamber 6-Sided Direct heating. Ideal for the culturing of non-adherent cells, the Incu-Shaker CO2 Mini combines a precision CO2 incubation chamber with an integral orbital shaker, providing the optimum conditions for suspension cell culture. With our unique and proprietary heat distribution technology, the incubator provides the highest levels of temperature accuracy with minimal fluctuation (within 0.1 ° C). The six-side heating system in combination with a ultra-low speed internal fan uniformly spread air throughout the stainless steel chamber without air exchange from the external environment. This minimal air flow design, significantly reduces the concerns of sample to sample contamination and allows for disabling of the fan for applications requiring solely mechanical convection. A powerful, beltless motor drive provides a horizontally circular shaking motion at a consistent