Learn More
Sigma Aldrich Fine Chemicals Biosciences Incu ShakerTM 10 L AC/DC i1EA
Catalog No. 502908584
Encompass
Incu-Shaker(TM) 10 L is a heavy-duty performer designed for heavy workloads and continuous use. Its outstanding temperature accuracy and uniformity are supported by proprietary heat distribution technology that ensures that temperatures are precisely maintained throughout the chamber.Inconsistencies due to stratification are reduced to negligible levels and the selected temperature is precisely maintained within 0.25% even in the corners.The powerful brushless induction drive is designed for years of trouble-free service even under continuous use conditions with heavy loads. Noise and vibration are minimized by an advanced computer designed motor isolation system.A standard rubber mat platform is included and is ideal for use with tissue culture flasks Petri dishes staining trays and other flat vessels. The convenient MAGic Clamp(TM) platform is also available for use with Erlenmeyer flasks and test tube racks.The unique magnetic attachment method is the easiest way to instantl
Catalog No. 50-290-8584 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7656861EA
Add to Cart
Add to Cart
Request a Quote
Incu-Shaker(TM) 10 L is a heavy-duty performer designed for heavy workloads and continuous use. Its outstanding temperature accuracy and uniformity are supported by proprietary heat distribution technology that ensures that temperatures are precisely maintained throughout the chamber.Inconsistencies due to stratification are reduced to negligible levels and the selected temperature is precisely maintained within 0.25% even in the corners.The powerful brushless induction drive is designed for years of trouble-free service even under continuous use conditions with heavy loads. Noise and vibration are minimized by an advanced computer designed motor isolation system.A standard rubber mat platform is included and is ideal for use with tissue culture flasks Petri dishes staining trays and other flat vessels. The convenient MAGic Clamp(TM) platform is also available for use with Erlenmeyer flasks and test tube racks.The unique magnetic attachment method is the easiest way to instantl