Learn More
Sigma Aldrich Fine Chemicals Biosciences Incu ShakerTM Mini CO2 wit1EA
Catalog No. 502908896
Encompass
Ideal for the culturing of non-adherent cells the Incu-Shaker CO2 Mini combines a precision CO2 incubation chamber with an intergal orbital shaker providing the optimum conditions for suspension cell culture.With our unique and proprietary heat distribution technology the incubator provides the highest levels of temperature accuracy with minimal fluctuation (within 0.1C). The six-side heating system in combination with a low speed internal fan uniformly spreads air throughout the stainless steel chamber (without air exchange from the external environment). For applications requiring solely gravity convection this internal fan can be disabledsignificantly reducing the concern of sample to sample contamination.A beltless motor drive provides a horizontally circular shaking motion at a consistent 19mm orbit. The included platform (9.5 x 11.5 in.) can be used for low speed shaking of a variety of laboratory vessels including Erlenmeyerflasks culture plates and media bottles. For hig
Catalog No. 50-290-8896 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7424451EA
Add to Cart
Add to Cart
Request a Quote
Ideal for the culturing of non-adherent cells the Incu-Shaker CO2 Mini combines a precision CO2 incubation chamber with an intergal orbital shaker providing the optimum conditions for suspension cell culture.With our unique and proprietary heat distribution technology the incubator provides the highest levels of temperature accuracy with minimal fluctuation (within 0.1C). The six-side heating system in combination with a low speed internal fan uniformly spreads air throughout the stainless steel chamber (without air exchange from the external environment). For applications requiring solely gravity convection this internal fan can be disabledsignificantly reducing the concern of sample to sample contamination.A beltless motor drive provides a horizontally circular shaking motion at a consistent 19mm orbit. The included platform (9.5 x 11.5 in.) can be used for low speed shaking of a variety of laboratory vessels including Erlenmeyerflasks culture plates and media bottles. For hig