Learn More
Sigma Aldrich Fine Chemicals Biosciences Incu ShakerTM Mini with no1EA
Catalog No. 502908603
Encompass
The Incu-Shaker Mini is the most compact shaking incubator in its class. Despite the remarkably small footprint a large 11.5 x 9.5 in. work space accepts several platforms and a variety of flasks tubes and other common vessels.The standard platform (included) features a non-slip rubber coated surface ideal for tissue culture flasks Petri dishes and staining trays..nbspExtremely compact just 11 in. wide.nbspExceptional temperature uniformity.nbspHorizontally circular 19 mm orbit for aeration mixing.nbspInstantly exchange flask clamps with MAGic Clamp(TM) accessories
Catalog No. 50-290-8603 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7635431EA
The Incu-Shaker Mini is the most compact shaking incubator in its class. Despite the remarkably small footprint a large 11.5 x 9.5 in. work space accepts several platforms and a variety of flasks tubes and other common vessels.The standard platform (included) features a non-slip rubber coated surface ideal for tissue culture flasks Petri dishes and staining trays..nbspExtremely compact just 11 in. wide.nbspExceptional temperature uniformity.nbspHorizontally circular 19 mm orbit for aeration mixing.nbspInstantly exchange flask clamps with MAGic Clamp(TM) accessories