Learn More
Grainger Scientific Industries Enviro-Genie Multi-Function Incubator, Refrigerated, 4°C to 75°C, 120 V
Catalog No. 502769227
Encompass_Preferred
The multi-function benchtop system integrates a refrigerator, incubator, rotator, rocker, and magnetic stirrer for the precise cultivation of microorganisms and tissue cells. Utilizing thermo-electric (Peltier) technology, the unit provides uniform temperature control without a compressor. It features a shatter-resistant viewing window for sample monitoring and an RS-232 interface for data acquisition. Digital controls allow for accurate, reproducible conditions across all functions.
  • Temperature range: 4°C to 75°C with 0.1°C resolution
  • Programmable 99-hour timer with audible and visible alarms
  • Dual integrated magnetic stirrers with speeds of 1 to 1,000 RPM
  • Rotation and rocking capabilities with speeds up to 35 RPM and 70 cycles per minute
  • Includes magnetic platform, non-skid tray, stackable shelf, bag mounting strips, and clip plates
Catalog No. 50-276-9227 Supplier Grainger Supplier No. 13R323
Add to Cart
Add to Cart
Request a Quote
The multi-function benchtop system integrates a refrigerator, incubator, rotator, rocker, and magnetic stirrer for the precise cultivation of microorganisms and tissue cells. Utilizing thermo-electric (Peltier) technology, the unit provides uniform temperature control without a compressor. It features a shatter-resistant viewing window for sample monitoring and an RS-232 interface for data acquisition. Digital controls allow for accurate, reproducible conditions across all functions.
  • Temperature range: 4°C to 75°C with 0.1°C resolution
  • Programmable 99-hour timer with audible and visible alarms
  • Dual integrated magnetic stirrers with speeds of 1 to 1,000 RPM
  • Rotation and rocking capabilities with speeds up to 35 RPM and 70 cycles per minute
  • Includes magnetic platform, non-skid tray, stackable shelf, bag mounting strips, and clip plates