Learn More
Grainger SHELLAB SSI5 Shaker Incubator, Digital, 120V AC, 1 ea
Catalog No. 502772492
Encompass_Preferred
This laboratory incubator features an integrated shaker designed for precise temperature control and sample agitation. Suitable for various research applications, it offers reliable performance with digital controls in a durable, floor-standing unit.
  • Temperature range of +8°C to +60°C
  • Digital control system
  • 10.82 cu. ft. capacity
  • Includes 2 shelves
  • 120V AC power requirement with 850 W power usage
  • Overall dimensions of 28 1/2 in. W x 29 1/2 in. D x 40 1/2 in. H
  • Chamber dimensions of 19 in. W x 20 1/2 in. D x 22 1/2 in. H
Catalog No. 50-277-2492 Supplier Grainger Supplier No. 13T093
Add to Cart
Add to Cart
Request a Quote
This laboratory incubator features an integrated shaker designed for precise temperature control and sample agitation. Suitable for various research applications, it offers reliable performance with digital controls in a durable, floor-standing unit.
  • Temperature range of +8°C to +60°C
  • Digital control system
  • 10.82 cu. ft. capacity
  • Includes 2 shelves
  • 120V AC power requirement with 850 W power usage
  • Overall dimensions of 28 1/2 in. W x 29 1/2 in. D x 40 1/2 in. H
  • Chamber dimensions of 19 in. W x 20 1/2 in. D x 22 1/2 in. H