Learn More
Biotang Inc Digital Incubating microplate shaker(2-plate) is a high performance microplate incubator and orbital shaker which accommodates up to four microplates, from 8deg.C above ambient to 70deg.C, from 200 to 1200 rpm, all parameters digital setting.
Catalog No. 507514966 Shop All BIOTANG Inc Products
Encompass_Preferred
Digital Incubating microplate shaker(2-plate) is a high performance microplate incubator and orbital shaker which accommodates up to four microplates, from 8deg.C above ambient to 70deg.C, from 200 to 1200 rpm, all parameters digital setting.
Catalog No. 50-751-4966 Supplier Biotang Inc Supplier No. MB1002A
Digital Incubating microplate shaker(2-plate) is a high performance microplate incubator and orbital shaker which accommodates up to four microplates, from 8deg.C above ambient to 70deg.C, from 200 to 1200 rpm, all parameters digital setting.