Learn More
Biotang Inc Digital Incubating microplate shaker(4-plate) is a high performance microplate incubator and orbital shaker which accommodates up to four microplates, from 8deg.C above ambient to 60deg.C, from 200 to 1200 rpm, all parameters digital setting.
Catalog No. 507514967 Shop All BIOTANG Inc Products
Encompass_Preferred
Digital Incubating microplate shaker(4-plate) is a high performance microplate incubator and orbital shaker which accommodates up to four microplates, from 8deg.C above ambient to 60deg.C, from 200 to 1200 rpm, all parameters digital setting.
Catalog No. 50-751-4967 Supplier Biotang Inc Supplier No. MB1004A
Digital Incubating microplate shaker(4-plate) is a high performance microplate incubator and orbital shaker which accommodates up to four microplates, from 8deg.C above ambient to 60deg.C, from 200 to 1200 rpm, all parameters digital setting.