Learn More
Biotang Inc Thermal Shaker for Microtubes, Digital Incubating (Cooling) Shaker, LCD display. Accurately control and display time, temperature and speed Temp.: 0~100; Speed200- 1500 rpm;Timer: 1-99h59min.
Catalog No. 507514969 Shop All BIOTANG Inc Products
Encompass_Preferred
Thermal Shaker for Microtubes, Digital Incubating (Cooling) Shaker, LCD display. Accurately control and display time, temperature and speed Temp.: 0~100; Speed200- 1500 rpm;Timer: 1-99h59min
Catalog No. 50-751-4969 Supplier Biotang Inc Supplier No. MSC100
Thermal Shaker for Microtubes, Digital Incubating (Cooling) Shaker, LCD display. Accurately control and display time, temperature and speed Temp.: 0~100; Speed200- 1500 rpm;Timer: 1-99h59min