Learn More
Biotang Inc Thermal Shaker for Microtubes, Digital Incubating Microplate Shaker, LCD display. Accurately control and display time, temperature and speed Temp.: RT.+5~100; Speed200- 1500 rpm;Timer: 1-99h59min, 10 selective blocks for tubes and plates.
Supplier:  Biotang Inc MS100
Encompass_Preferred
Thermal Shaker for Microtubes, Digital Incubating Microplate Shaker, LCD display. Accurately control and display time, temperature and speed Temp.: RT.+5~100; Speed200- 1500 rpm;Timer: 1-99h59min, 10 selective blocks for tubes and plates.
Catalog No. 50-751-4968