Learn More
Andwin Scientific INCUBATING ORBITL SHAKER 3500I
SDP
Catalog No. NC2118843
Encompass_Preferred
VWR Incubating Orbital Shaker, Model 3500IThe microprocessor control of the Model 3500I provides consistent, uniform mixing action.Microprocessor control with digital display for speed, temperature, and timeOpti-flow forced ventilation systemTriple eccentric drive with brushless motorFive-year warrantyNIST-Traceable, ISO/IEC 17025 accredited calibration certificate from Troemner availableA spill-resistant touchpad adjusts the temperature in 1 C increments
This item is not returnable. View return policy
Catalog No. NC2118843 Supplier Andwin Scientific Supplier No. 12620946
Add to Cart
Add to Cart
Request a Quote
VWR Incubating Orbital Shaker, Model 3500IThe microprocessor control of the Model 3500I provides consistent, uniform mixing action.Microprocessor control with digital display for speed, temperature, and timeOpti-flow forced ventilation systemTriple eccentric drive with brushless motorFive-year warrantyNIST-Traceable, ISO/IEC 17025 accredited calibration certificate from Troemner availableA spill-resistant touchpad adjusts the temperature in 1 C increments