Learn More
Electron Microscopy Sciences Buffer Solution with long term stability, has a buffer additive that prevents aspecific binding of antibodies and immunodetection reagents to polycationic sites in the specimen.
SDP
Catalog No. NC3245924
Encompass_Preferred
Ready-to-Use Incubation Buffer Solution with long term stability, has an unique buffer additive (BSA-c™) that prevents aspecific binding of antibodies and immunodetection reagents to polycationic sites in the specimen, such as histone proteins in the nucleus.
Catalog No. NC3245924 Supplier Electron Microscopy Sciences Supplier No. 25561
Ready-to-Use Incubation Buffer Solution with long term stability, has an unique buffer additive (BSA-c™) that prevents aspecific binding of antibodies and immunodetection reagents to polycationic sites in the specimen, such as histone proteins in the nucleus.