Learn More
Electron Microscopy Sciences CoverWell™ Incubation Chambers, 500 mcL, 0.2 mm depth
SDP
Catalog No. 5025149
Encompass_Preferred
CoverWell incubation reusable chambers are reusable, easy to apply incubation chambers that attach to slides without the use of adhesive.
Microwavable for permeabilization and retrieval steps.
Preserve kinetic (non-capillary) fluid dynamics for better reagent mixing and lower backgrounds.
Eliminate precipitate deposits on specimens by incubating slides and specimens upside down during enzymatic color precipitation reaction.
Provides an exceptionally secure seal during submerged water bath and/or high temperature incubations.
Rnase and Dnase free, hydrophobic
Easily removed (reusable)
Adheres to wet or dry surfaces.
Volume: 500 µl
Depth: 0.2 mm
Catalog No. 50-251-49 Supplier Electron Microscopy Sciences Supplier No. 7032450
CoverWell incubation reusable chambers are reusable, easy to apply incubation chambers that attach to slides without the use of adhesive.
Microwavable for permeabilization and retrieval steps.
Preserve kinetic (non-capillary) fluid dynamics for better reagent mixing and lower backgrounds.
Eliminate precipitate deposits on specimens by incubating slides and specimens upside down during enzymatic color precipitation reaction.
Provides an exceptionally secure seal during submerged water bath and/or high temperature incubations.
Rnase and Dnase free, hydrophobic
Easily removed (reusable)
Adheres to wet or dry surfaces.
Volume: 500 µl
Depth: 0.2 mm