Learn More
Chemglass Life Sciences Incubator, 5.0 Cu. Ft., Water Jacketed, CO², 120V
SDP
Supplier:  Chemglass Life Sciences CLS2447612
Encompass
Anti-Corrosion Anode. Patented Copper-Housed HEPA Filter for Superior Contamination Prevention. Factory Pre-Set, Door Thermostat. Pre-Heated CO2 Gas Inlet for Increased Temperature Uniformity. Front-Mounted Connections for Convenience. Over Temperature and CO2 Alarms. Inner Glass Viewing Door. Stainless Steel Chamber and Autoclavable Door Gasket Make Cleaning Easy. 5-Year Limited Warranty. The CLS-2447 series of CO2 incubators offers dependable infrared (IR), CO2 sensor control and is ideal for QA / QC applications. The incubators include an anti-corrosion anode which allows the use of distilled or tap water in the compact water jacket. De-ionized water should not be used. Anti-contamination features make cleaning a breeze and the CO2 sample port is located on the front control panel for quick and easy access. Incubators have microprocessor controllers. The humidity level inside the chamber is maintained at approximately 95%.
Catalog No. 50-143-552
Add to Cart
Request a Quote