Learn More
Chemglass Life Sciences Incubator, 10L, Incu-Shake, with Non-Slip Rubber Mat and Touch Screen, 120V
SDP
Catalog No. 50143351
Encompass
The Incu-Shaker Mini is the most compact shaking incubator in its class. Despite the remarkably small footprint, a large 11. 5 x 9. 5 in. work space accepts several platforms and a variety of flasks, tubes and other common vessels.  The standard platform (included) features a non-slip, rubber coated surface, ideal for tissue culture flasks, Petri dishes and staining trays.  The convenient MAGic Clamp platform is also available for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attach- ment method is the easiest way to instantly change between flask clamps of different sizes. Extremely compact, just 11" wide. TOUCH Screen control over time, speed and temperature. Horizontally circular 19mm orbit for aeration and mixing. Instantly exchange flask clamps with MAGic Clamp accessories (PATENTED). Program STEP feature for linking consecutive programs.
Catalog No. 50-143-351 Supplier Chemglass Life Sciences Supplier No. CLS2543002
Add to Cart
Add to Cart
Request a Quote
The Incu-Shaker Mini is the most compact shaking incubator in its class. Despite the remarkably small footprint, a large 11. 5 x 9. 5 in. work space accepts several platforms and a variety of flasks, tubes and other common vessels.  The standard platform (included) features a non-slip, rubber coated surface, ideal for tissue culture flasks, Petri dishes and staining trays.  The convenient MAGic Clamp platform is also available for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attach- ment method is the easiest way to instantly change between flask clamps of different sizes. Extremely compact, just 11" wide. TOUCH Screen control over time, speed and temperature. Horizontally circular 19mm orbit for aeration and mixing. Instantly exchange flask clamps with MAGic Clamp accessories (PATENTED). Program STEP feature for linking consecutive programs.