Learn More
Chemglass Life Sciences Incubator, 10L, Incu-Shake, Refrigerated, with Non-Slip Rubber Mat, 120V
SDP
Catalog No. 501311306
Encompass
Incubator, 10L, Incu-Shake, Refrigerated, with Non-Slip Rubber Mat, 120V. The new Incu-Shaker™ series of shaking incubators provide digitally regulated temperature with precisely controlled circular (orbital) mixing, optimized for the culture of a wide variety of cell types.The 10L Incu-Shakers™ are equipped with a large 18 x 18 inch platform for high capacity applications.The standard platform (included with both models) features a non-slip, rubber coated surface, ideal for tissue culture flasks, Petri dishes and staining trays.Temperature, shaking speed and run time are displayed on the large LED control panel located on the front of the unit. The integral microprocessor includes a constant monitoring system that verifies and maintains accuracy through the duration of the program. Sophisticated over-temperature and over-speed controls ensure long life, safety and sample integrity.
Catalog No. 50-131-1306 Supplier Chemglass Life Sciences Supplier No. CLS25430R2
Add to Cart
Add to Cart
Request a Quote
Incubator, 10L, Incu-Shake, Refrigerated, with Non-Slip Rubber Mat, 120V. The new Incu-Shaker™ series of shaking incubators provide digitally regulated temperature with precisely controlled circular (orbital) mixing, optimized for the culture of a wide variety of cell types.The 10L Incu-Shakers™ are equipped with a large 18 x 18 inch platform for high capacity applications.The standard platform (included with both models) features a non-slip, rubber coated surface, ideal for tissue culture flasks, Petri dishes and staining trays.Temperature, shaking speed and run time are displayed on the large LED control panel located on the front of the unit. The integral microprocessor includes a constant monitoring system that verifies and maintains accuracy through the duration of the program. Sophisticated over-temperature and over-speed controls ensure long life, safety and sample integrity.