Learn More
Percival Scientific Biological Incubator, I66NL, 62.4 cu. ft., non-lighted, 6 tiers, 2-44° C ±0.5°, IntellusUltra controller
SDP
Supplier:  Percival Scientific I66NL
Encompass_Preferred
The I66NL incubator is frequently used for C. elegans, nematodes, yeast/fungi, insects, bacteria and BOD. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Twelve white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. For additional options contact sales@percival-scientific.com for a free consultation.
Catalog No. NC2948053
Only null left
Add to Cart
Request a Quote