Learn More
Grainger Shel Lab CO2 Incubator, Water Jacketed, 1.5 cu.ft., IR, HEPA, 1 ea
Catalog No. 502773145
Encompass_Preferred
This water-jacketed CO2 incubator is designed for professional cell culture applications, featuring advanced PID microprocessor control for precise environmental stability. Its 300 series stainless steel interior with coved corners facilitates easy cleaning and decontamination.
  • Advanced PID control ensures rapid recovery and precise stability
  • Infrared (IR) sensor provides quick CO2 recovery following door openings
  • Heated door minimizes condensation and enhances temperature uniformity
  • Patented copper-caged HEPA filter captures 99.97% of airborne microbes at 0.3 microns
  • Exclusive heated copper CO2 inlet prevents moisture introduction
  • Autoclavable stainless steel shelving system supports flexible interior configuration
  • Includes USB data logging capabilities
Catalog No. 50-277-3145 Supplier Grainger Supplier No. 13T082
Add to Cart
Add to Cart
Request a Quote
This water-jacketed CO2 incubator is designed for professional cell culture applications, featuring advanced PID microprocessor control for precise environmental stability. Its 300 series stainless steel interior with coved corners facilitates easy cleaning and decontamination.
  • Advanced PID control ensures rapid recovery and precise stability
  • Infrared (IR) sensor provides quick CO2 recovery following door openings
  • Heated door minimizes condensation and enhances temperature uniformity
  • Patented copper-caged HEPA filter captures 99.97% of airborne microbes at 0.3 microns
  • Exclusive heated copper CO2 inlet prevents moisture introduction
  • Autoclavable stainless steel shelving system supports flexible interior configuration
  • Includes USB data logging capabilities