Learn More
Grainger SHEL LAB Incubator, SCO5A, Air Jacket, 5 cu ft, 1 ea
Catalog No. 502772488
Encompass_Preferred
This air jacketed CO2 incubator provides a stable environment for cell and tissue culture applications. Designed with a 5 cu. ft. capacity and an internal stainless steel chamber, it offers precise regulation of temperature, humidity, and carbon dioxide levels to support optimal cell growth.
  • Features digital controls and a digital display for accurate monitoring.
  • Includes an integrated audio alarm system for safety.
  • Constructed with a durable steel housing and a swing glass door.
  • Equipped with three stainless steel shelves.
  • Includes a HEPA filter for clean air circulation.
  • Features a 4 in. side-mounted probe port.
  • Operates at a temperature range of ambient +5 °C to 60 °C with an accuracy tolerance of ±0.25 °C at 37 °C.
  • Supports CO2 concentration from 1% to 20% and up to 95% humidity.
Catalog No. 50-277-2488 Supplier Grainger Supplier No. 13T085
Add to Cart
Add to Cart
Request a Quote
This air jacketed CO2 incubator provides a stable environment for cell and tissue culture applications. Designed with a 5 cu. ft. capacity and an internal stainless steel chamber, it offers precise regulation of temperature, humidity, and carbon dioxide levels to support optimal cell growth.
  • Features digital controls and a digital display for accurate monitoring.
  • Includes an integrated audio alarm system for safety.
  • Constructed with a durable steel housing and a swing glass door.
  • Equipped with three stainless steel shelves.
  • Includes a HEPA filter for clean air circulation.
  • Features a 4 in. side-mounted probe port.
  • Operates at a temperature range of ambient +5 °C to 60 °C with an accuracy tolerance of ±0.25 °C at 37 °C.
  • Supports CO2 concentration from 1% to 20% and up to 95% humidity.