Learn More
Med Vet International McKesson STER-ALL Performance Incubator, 73-NDB060, Each
Catalog No. 503044484
Encompass_Preferred
Dry Block Incubator McKesson 15 Wells, 11 mm With vial crusher Pre-set at 60°C for operation on 120 VAC 60 HZ cycle AC Temperature is calibrated using a thermometer found to be within the National Institute of Standards and Technology Not made with natural rubber latex Packaged: Each
Catalog No. 50-304-4484 Supplier Med Vet International Supplier No. 524872EA
Dry Block Incubator McKesson 15 Wells, 11 mm With vial crusher Pre-set at 60°C for operation on 120 VAC 60 HZ cycle AC Temperature is calibrated using a thermometer found to be within the National Institute of Standards and Technology Not made with natural rubber latex Packaged: Each