Learn More
Chemglass Life Sciences Incubator, 58 Cu. Ft., Large Capacity, CO², 120V
SDP
Supplier:  Chemglass Life Sciences CLS2449060
Encompass
Microprocessor Temperature Control. All Stainless Steel Interior. LED Display of Set point and Chamber Temperature. Infrared CO2 Sensor. Interior Electrical Outlets. 24-Month Limited Warranty. The 2449 series incubators are well-suited for roller bottle apparatus and high-volume tissue culture applications and are ideal for cell harvesting. The 28 and 40 cu. ft. models are suited for roller bottle apparatus. These large capacity incubators maximize laboratory space in a convenient floor model design. The incubators are supplied with (4) one amp interior electrical outlets and gentle mechanical air convection that ensures excellent temperature uniformity and eliminates cold spots. An infrared system accurately controls CO2 levels, provides fast CO2 recovery after door openings and is not affected by temperature or humidity. This unit is supplied with six stainless steel re-enforced shelves, which are adjustable on 1/2 inch increments.
Catalog No. 50-143-514
Add to Cart
Request a Quote