Learn More
Cole-Parmer Stuart Analog Incubator, acrylic, 120 V
Supplier:  Cole-Parmer 0111000
Encompass_Preferred
Clear acrylic top and sides make it easy to view your sample from all sides—ideal for plant propagation, simulation of tropical conditions, and corrosion testing. These incubators do not have a base so they can be placed directly over complete instruments; order base separately from "Accessories". Forced air circulation and electronic temperature control ensure accurately maintained conditions. Select from analog or digital models. A simple calibrated knob lets you set the temperature on the analog model. Digital model with LCD lets you set and view chamber temperature. Both models have a push switch preset at 37°C. Safety feature cuts off heat at 72°C.
Catalog No. 50-196-0402
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.