Learn More
Chemglass Life Sciences Incubator, Peltier-Cooled, B.O.D., 19.3 Cu Ft, 120V
SDP
Catalog No. 50143494
Encompass
Our B. O. D. Thermoelectric Cooled Incubator uses innovative thermoelectric cooling technology that eliminates the need for a refrigeration compressor. This means these units use 78% less power than alternative models and reduce room air conditioning loads by 75%. They also include 75-pound capacity shelves, which eliminates sagging. These incubators meet APHA specifications for Biochemical Oxygen Demand (B. O. D. ) and include a mechanical convection system to ensure even air distribution, digital temperature set controller, over temperature limit control, and a digital temperature display. A one amp interior outlet allows use of shakers, stirrers, roller bottles or other apparatus. Units are supplied with adjustable leveling feet. Low temperature incubators are ideal for testing of sewage and wastewater, as well as other low and room temperature applications. 24 Month Limited Warranty
Catalog No. 50-143-494 Supplier Chemglass Life Sciences Supplier No. CLS2442G03
Add to Cart
Add to Cart
Request a Quote
Our B. O. D. Thermoelectric Cooled Incubator uses innovative thermoelectric cooling technology that eliminates the need for a refrigeration compressor. This means these units use 78% less power than alternative models and reduce room air conditioning loads by 75%. They also include 75-pound capacity shelves, which eliminates sagging. These incubators meet APHA specifications for Biochemical Oxygen Demand (B. O. D. ) and include a mechanical convection system to ensure even air distribution, digital temperature set controller, over temperature limit control, and a digital temperature display. A one amp interior outlet allows use of shakers, stirrers, roller bottles or other apparatus. Units are supplied with adjustable leveling feet. Low temperature incubators are ideal for testing of sewage and wastewater, as well as other low and room temperature applications. 24 Month Limited Warranty