Learn More
Grainger SHEL LAB SCO5W Water Jacket CO2 Incubator, 5.1 cu. ft., Stainless Steel, 1 ea
Catalog No. 502772431
Encompass_Preferred
The water jacketed carbon dioxide incubator provides a stable environment for sensitive cell cultures. This laboratory incubator features a robust stainless steel chamber designed to maintain precise atmospheric conditions and temperature uniformity for optimal cell growth.
  • Water jacket convection for superior temperature stability
  • Infrared sensor for accurate CO2 control
  • Integrated HEPA filter system to minimize contamination risks
  • Digital display and intuitive control interface
  • Includes three adjustable stainless steel shelves
  • Audio alarm system for monitoring key parameters
  • Side access port for easy monitoring and instrumentation
Catalog No. 50-277-2431 Supplier Grainger Supplier No. 13T083
Add to Cart
Add to Cart
Request a Quote
The water jacketed carbon dioxide incubator provides a stable environment for sensitive cell cultures. This laboratory incubator features a robust stainless steel chamber designed to maintain precise atmospheric conditions and temperature uniformity for optimal cell growth.
  • Water jacket convection for superior temperature stability
  • Infrared sensor for accurate CO2 control
  • Integrated HEPA filter system to minimize contamination risks
  • Digital display and intuitive control interface
  • Includes three adjustable stainless steel shelves
  • Audio alarm system for monitoring key parameters
  • Side access port for easy monitoring and instrumentation