Learn More
Grainger SHEL LAB CO2 Incubator, Water Jacketed, 167 L, Digital
Catalog No. 502769241
Encompass_Preferred
This water-jacketed CO2 incubator provides a stable and precise environment for sensitive cell culture applications. Designed with an advanced PID microprocessor for reliable temperature and gas control, this unit features a durable stainless steel interior and a water-jacketed heating system to maintain uniform conditions.
  • Water jacketed design for thermal stability
  • Advanced PID microprocessor control
  • Infrared (IR) CO2 sensor for accurate gas monitoring
  • Audible alarm system included
  • Stainless steel interior for corrosion resistance
  • Includes probe port and complete accessory kit
  • Digital display for easy monitoring of set parameters
Catalog No. 50-276-9241 Supplier Grainger Supplier No. 13T080
Add to Cart
Add to Cart
Request a Quote
This water-jacketed CO2 incubator provides a stable and precise environment for sensitive cell culture applications. Designed with an advanced PID microprocessor for reliable temperature and gas control, this unit features a durable stainless steel interior and a water-jacketed heating system to maintain uniform conditions.
  • Water jacketed design for thermal stability
  • Advanced PID microprocessor control
  • Infrared (IR) CO2 sensor for accurate gas monitoring
  • Audible alarm system included
  • Stainless steel interior for corrosion resistance
  • Includes probe port and complete accessory kit
  • Digital display for easy monitoring of set parameters