Learn More
Hach Company Incubator, Culture, Low-Profile 110V
Catalog No. 501996373 Shop All Hach Company Products
Encompass
Hach's Low-Profile Culture Model 153 Incubator exceeds performance criteria for Standard Methods, EPA, & ASTM microbiological methods of an accuracy of ±0.1 °C @ 37 °C. Five heating elements minimize "hot spots" and contribue to a uniformity of ±0.25 °C @ 37 °C. Features a High Limit Thermostat and Warning Light to guard against overheating during use, stainless steel interior and shelves (2), and maintenance free heating system. Improved inner viewing door allows you to check on your samples without large interior temperature swings.Capacity: 6.67 ft³ Temperature Range: 5° above Ambient to 70 °C (Accuracy: ±0.1°C @ 20°C) External Dimensions (W x D x H): 762 x 762 x 813 mm (30 x 30 x 32")
Catalog No. 50-199-6373 Supplier Hach Company Supplier No. 2619200
Hach's Low-Profile Culture Model 153 Incubator exceeds performance criteria for Standard Methods, EPA, & ASTM microbiological methods of an accuracy of ±0.1 °C @ 37 °C. Five heating elements minimize "hot spots" and contribue to a uniformity of ±0.25 °C @ 37 °C. Features a High Limit Thermostat and Warning Light to guard against overheating during use, stainless steel interior and shelves (2), and maintenance free heating system. Improved inner viewing door allows you to check on your samples without large interior temperature swings.Capacity: 6.67 ft³ Temperature Range: 5° above Ambient to 70 °C (Accuracy: ±0.1°C @ 20°C) External Dimensions (W x D x H): 762 x 762 x 813 mm (30 x 30 x 32")