Learn More
Chemglass Life Sciences Incubator, Digital, 115V, myTemp Mini CO2
SDP
Catalog No. 5000083206
Encompass
Digital Control Compact Heats and Cools Infrared CO2 Sensor. At only 13 inches wide, the myTemp Mini CO2 is the smallest and most economical incubator for applications requiring CO2 gas control. Its small footprint makes the incubator ideal for use inside of biological safety cabinets, isolating sensitive cultures or for any laboratory simply trying to conserve bench space. With a thick layer of high efficiency insulation surrounding the chamber, the incubator is also ideal for transportation of cells. Once power has been disconnected, the incubator maintains temperature levels within 10% of the set parameter for up to 1 hour*. In addition each model includes a 12V vehicle power adapter for operating the instrument in a car, boat, etc. (* If disconnected from the CO2 supply, CO2 percentage decreases by approx. 1%/30 min). Both temperature and CO2 percentage are digitally adjustable and displayed in real time on the large LED control panel. This panel also features a "Cal" function, for
Catalog No. 50-000-83206 Supplier Chemglass Life Sciences Supplier No. CLS2457H01
Digital Control Compact Heats and Cools Infrared CO2 Sensor. At only 13 inches wide, the myTemp Mini CO2 is the smallest and most economical incubator for applications requiring CO2 gas control. Its small footprint makes the incubator ideal for use inside of biological safety cabinets, isolating sensitive cultures or for any laboratory simply trying to conserve bench space. With a thick layer of high efficiency insulation surrounding the chamber, the incubator is also ideal for transportation of cells. Once power has been disconnected, the incubator maintains temperature levels within 10% of the set parameter for up to 1 hour*. In addition each model includes a 12V vehicle power adapter for operating the instrument in a car, boat, etc. (* If disconnected from the CO2 supply, CO2 percentage decreases by approx. 1%/30 min). Both temperature and CO2 percentage are digitally adjustable and displayed in real time on the large LED control panel. This panel also features a "Cal" function, for